Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007527.1 | NP_001007528.1 | 2 - 3 | 580 | 189 |
XM_011514162.2 | XP_011512464.1 | 2 - 3 | 1022 | 189 | |
XM_017010024.1 | XP_016865513.1 | 2 - 3 | 526 | 189 | |
GenBank mRNA | AK091295.1 | - | 2 - 3 | 520 | 189 |
AK300433.1 | - | 3 - 4 | 490 | 103 | |
AK307575.1 | - | 2 - 3 | 526 | 189 | |
BC110506.2 | - | 2 - 3 | 519 | 189 | |
BX647340.1 | - | 2 - 3 | 555 | 189 | |
CR749399.1 | - | 2 - 3 | 580 | 189 |