Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033549.2 | NP_001028721.1 | 8 - 9 | 979 | 70 |
NM_001288756.1 | NP_001275685.1 | 8 - 9 | 930 | 70 | |
NM_001288757.1 | NP_001275686.1 | 5 - 6 | 754 | 70 | |
NM_014173.3 | NP_054892.2 | 8 - 9 | 941 | 70 | |
GenBank mRNA | AK000578.1 | - | 8 - 9 | 856 | 70 |
AK025970.1 | - | 8 - 9 | 874 | 70 | |
AK225047.1 | - | 8 - 9 | 863 | 70 | |
AL136692.1 | - | 8 - 9 | 873 | 70 | |
BC000788.2 | - | 8 - 9 | 840 | 70 | |
BC006244.1 | - | 8 - 9 | 881 | 70 | |
BC091491.1 | - | 8 - 9 | 887 | 70 | |
CR533526.1 | - | 7 - 8 | 776 | 70 | |
CX866662.1 | - | 5 - 6 | 627 | 70 |