Assay Design



Gene Transcripts

Gene Symbol
SYT17
Entrez Gene ID
51760
Gene Name
synaptotagmin 17
Gene Aliases
sytXVII
Chromosome Location
Chr.16: 19167834 - 19268334 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016524.3 NP_057608.2 2 - 3 464 65
XM_017023280.1 XP_016878769.1 2 - 3 458 65
GenBank mRNA BC004518.2 - 2 - 3 40 65
CA389452.1 - 2 - 3 361 65

View Full Product Details