Assay Design



Gene Transcripts

Gene Symbol
BLCAP
Entrez Gene ID
10904
Gene Name
bladder cancer associated protein
Gene Aliases
BC10
Chromosome Location
Chr.20: 37517417 - 37527931 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006698.3 NP_006689.1 1 - 2 137 128
GenBank mRNA CB155706.1 - 1 - 2 107 128
DA262965.1 - 1 - 2 137 128

View Full Product Details