Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317931.1 | NP_001304860.1 | 2 - 3 | 505 | 50 |
NM_003116.2 | NP_003107.1 | 2 - 3 | 524 | 50 | |
XM_005260520.4 | XP_005260577.1 | 2 - 3 | 170 | 50 | |
XM_011529009.2 | XP_011527311.1 | 2 - 3 | 420 | 50 | |
GenBank mRNA | AF262992.1 | - | 2 - 3 | 521 | 50 |
BC143783.1 | - | 2 - 3 | 426 | 50 |