Assay Design



Gene Transcripts

Gene Symbol
TBC1D2B
Entrez Gene ID
23102
Gene Name
TBC1 domain family member 2B
Gene Aliases
-
Chromosome Location
Chr.15: 77994985 - 78077708 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144572.1 NP_653173.1 12 - 13 2697 64
XM_011521387.2 XP_011519689.1 11 - 12 2642 64
XM_017022022.1 XP_016877511.1 14 - 15 2718 64
GenBank mRNA BC033712.1 - 7 - 8 1480 64
BC140766.1 - 11 - 12 2342 64
BC144482.1 - 11 - 12 2342 64

View Full Product Details