Assay Design



Gene Transcripts

Gene Symbol
TBC1D2B
Entrez Gene ID
23102
Gene Name
TBC1 domain family member 2B
Gene Aliases
-
Chromosome Location
Chr.15: 77994985 - 78077708 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015079.5 NP_055894.6 7 - 8 1575 70
NM_144572.1 NP_653173.1 7 - 8 1575 70
XM_017022022.1 XP_016877511.1 9 - 10 1596 70
GenBank mRNA AB028978.1 - 7 - 8 1438 70
BC033712.1 - 2 - 3 358 70
BC140766.1 - 6 - 7 1220 70
BC144482.1 - 6 - 7 1220 70
BC152465.1 - 7 - 8 1438 70

View Full Product Details