Assay Design



Gene Transcripts

Gene Symbol
CBLN4
Entrez Gene ID
140689
Gene Name
cerebellin 4 precursor
Gene Aliases
CBLNL1
Chromosome Location
Chr.20: 55997357 - 56005472 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080617.5 NP_542184.1 2 - 3 1703 130
GenBank mRNA AK289640.1 - 2 - 3 1507 130
AY358527.1 - 2 - 3 738 130
BC050026.1 - 2 - 3 1705 130
BC069402.1 - 2 - 3 402 130
BC069488.1 - 2 - 3 402 130
BC069516.1 - 2 - 3 402 130

View Full Product Details