Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007036.4 | NP_008967.1 | 1 - 2 | 372 | 86 |
GenBank mRNA | AK311817.1 | - | 1 - 2 | 379 | 86 |
BC011989.1 | - | 1 - 2 | 362 | 86 | |
BM423977.1 | - | 1 - 2 | 449 | 86 | |
X89426.1 | - | 1 - 2 | 358 | 86 |