Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252226.1 | NP_001239155.1 | 13 - 14 | 1896 | 97 |
NM_006622.3 | NP_006613.2 | 12 - 13 | 1938 | 97 | |
GenBank mRNA | AF059617.1 | - | 12 - 13 | 2058 | 97 |
AF223574.1 | - | 12 - 13 | 1881 | 97 | |
AK027851.1 | - | 12 - 13 | 1882 | 97 | |
AK098163.1 | - | 11 - 12 | 1479 | 97 | |
AK312505.1 | - | 12 - 13 | 1886 | 97 | |
BC013879.2 | - | 12 - 13 | 1871 | 97 |