Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199922.1 | NP_001186851.1 | 9 - 10 | 1263 | 77 |
NM_170601.4 | NP_733746.1 | 7 - 8 | 1132 | 77 | |
XM_017017930.1 | XP_016873419.1 | 8 - 9 | 3652 | 77 | |
GenBank mRNA | AF300796.1 | - | 9 - 10 | 1263 | 77 |
AF303378.1 | - | 7 - 8 | 971 | 77 | |
AK056093.1 | - | 7 - 8 | 998 | 77 | |
AK092224.1 | - | 3 - 4 | 311 | 77 | |
AL137496.1 | - | 7 - 8 | 954 | 77 | |
BC022456.1 | - | 7 - 8 | 986 | 77 | |
BC068450.1 | - | 7 - 8 | 1118 | 77 |