Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199922.1 | NP_001186851.1 | 10 - 11 | 1429 | 83 |
NM_170601.4 | NP_733746.1 | 8 - 9 | 1298 | 83 | |
XM_017017930.1 | XP_016873419.1 | 9 - 10 | 3818 | 83 | |
GenBank mRNA | AF300796.1 | - | 10 - 11 | 1429 | 83 |
AF303378.1 | - | 8 - 9 | 1137 | 83 | |
AK056093.1 | - | 8 - 9 | 1164 | 83 | |
AK092224.1 | - | 4 - 5 | 477 | 83 | |
AL137496.1 | - | 8 - 9 | 1120 | 83 | |
BC022456.1 | - | 8 - 9 | 1152 | 83 | |
BC068450.1 | - | 8 - 9 | 1284 | 83 |