Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006469.4 | NP_006460.2 | 1 - 2 | 370 | 155 |
XM_005244843.3 | XP_005244900.1 | 1 - 2 | 326 | 155 | |
XM_011509079.2 | XP_011507381.1 | 1 - 2 | 380 | 155 | |
XM_011509080.2 | XP_011507382.1 | 1 - 2 | 384 | 155 | |
XM_017000084.1 | XP_016855573.1 | 1 - 2 | 326 | 155 | |
GenBank mRNA | AB020657.1 | - | 1 - 2 | 377 | 155 |
AF161553.1 | - | 1 - 2 | 286 | 155 | |
AF205218.1 | - | 1 - 2 | 302 | 155 | |
AJ012449.1 | - | 1 - 2 | 284 | 155 | |
AK023123.1 | - | 1 - 2 | 314 | 155 | |
AK292431.1 | - | 1 - 2 | 120 | 155 | |
BC067739.1 | - | 1 - 2 | 270 | 155 | |
DQ443528.1 | - | 1 - 2 | 259 | 155 |