Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160210.1 | NP_001153682.1 | 12 - 13 | 1429 | 75 |
NM_014251.2 | NP_055066.1 | 12 - 13 | 1426 | 75 | |
NR_027662.1 | 11 - 12 | 1310 | 75 | ||
XM_006715831.3 | XP_006715894.1 | 12 - 13 | 1362 | 75 | |
XM_011515727.2 | XP_011514029.1 | 13 - 14 | 1550 | 75 | |
XM_017011663.1 | XP_016867152.1 | 11 - 12 | 1277 | 75 | |
XM_017011664.1 | XP_016867153.1 | 11 - 12 | 1312 | 75 | |
XM_017011665.1 | XP_016867154.1 | 10 - 11 | 1189 | 75 | |
GenBank mRNA | AF118838.1 | - | 12 - 13 | 1372 | 75 |
AJ496569.1 | - | 12 - 13 | 1325 | 75 | |
AK000766.1 | - | 12 - 13 | 1345 | 75 | |
AK025227.1 | - | 9 - 10 | 941 | 75 | |
AK025779.1 | - | 7 - 8 | 782 | 75 | |
AK294629.1 | - | 11 - 12 | 1175 | 75 | |
BC006566.2 | - | 12 - 13 | 1308 | 75 | |
Y17571.1 | - | 12 - 13 | 1275 | 75 |