Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160210.1 | NP_001153682.1 | 15 - 16 | 1790 | 106 |
NM_014251.2 | NP_055066.1 | 15 - 16 | 1787 | 106 | |
NR_027662.1 | 14 - 15 | 1671 | 106 | ||
XM_006715831.3 | XP_006715894.1 | 15 - 16 | 1723 | 106 | |
XM_017011663.1 | XP_016867152.1 | 14 - 15 | 1638 | 106 | |
XM_017011664.1 | XP_016867153.1 | 14 - 15 | 1673 | 106 | |
XM_017011665.1 | XP_016867154.1 | 13 - 14 | 1550 | 106 | |
GenBank mRNA | AF118838.1 | - | 15 - 16 | 1733 | 106 |
AJ496569.1 | - | 15 - 16 | 1686 | 106 | |
AK000766.1 | - | 15 - 16 | 1706 | 106 | |
AK025227.1 | - | 12 - 13 | 1302 | 106 | |
AK025779.1 | - | 10 - 11 | 1143 | 106 | |
AK222864.1 | - | 15 - 16 | 1706 | 106 | |
AK294629.1 | - | 14 - 15 | 1536 | 106 | |
BC006566.2 | - | 15 - 16 | 1669 | 106 |