Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160210.1 | NP_001153682.1 | 7 - 8 | 938 | 78 |
NM_014251.2 | NP_055066.1 | 7 - 8 | 938 | 78 | |
NR_027662.1 | 6 - 7 | 822 | 78 | ||
XM_006715831.3 | XP_006715894.1 | 7 - 8 | 874 | 78 | |
XM_011515727.2 | XP_011514029.1 | 8 - 9 | 1062 | 78 | |
XM_017011663.1 | XP_016867152.1 | 6 - 7 | 789 | 78 | |
GenBank mRNA | AF118838.1 | - | 7 - 8 | 884 | 78 |
AJ496569.1 | - | 7 - 8 | 834 | 78 | |
AK000766.1 | - | 7 - 8 | 857 | 78 | |
AK025227.1 | - | 4 - 5 | 453 | 78 | |
AK025779.1 | - | 2 - 3 | 294 | 78 | |
AK222864.1 | - | 7 - 8 | 857 | 78 | |
AK294629.1 | - | 6 - 7 | 687 | 78 | |
BC006566.2 | - | 7 - 8 | 820 | 78 | |
Y17571.1 | - | 7 - 8 | 787 | 78 |