Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015395.2 | NP_056210.1 | 12 - 13 | 2123 | 67 |
XM_005250253.3 | XP_005250310.1 | 12 - 13 | 2095 | 67 | |
XM_017011937.1 | XP_016867426.1 | 11 - 12 | 2005 | 67 | |
GenBank mRNA | AB037779.1 | - | 10 - 11 | 1699 | 67 |
AK057048.1 | - | 11 - 12 | 1850 | 67 | |
AK292996.1 | - | 12 - 13 | 2025 | 67 | |
AL117495.1 | - | 1 - 2 | 1021 | 67 | |
BC053591.1 | - | 12 - 13 | 2028 | 67 |