Assay Design



Gene Transcripts

Gene Symbol
SLC22A16
Entrez Gene ID
85413
Gene Name
solute carrier family 22 member 16
Gene Aliases
CT2, FLIPT2, HEL-S-18, OAT6, OCT6, OKB1, dJ261K5.1
Chromosome Location
Chr.6: 110424587 - 110476641 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033125.3 NP_149116.2 1 - 2 119 115
XM_011536205.2 XP_011534507.1 1 - 2 91 115
XM_011536206.2 XP_011534508.1 1 - 2 103 115
XM_011536208.2 XP_011534510.1 1 - 2 103 115
XM_011536210.1 XP_011534512.1 1 - 2 91 115
XM_011536211.2 XP_011534513.1 1 - 2 103 115
GenBank mRNA AF268892.1 - 1 - 2 52 115
AK304546.1 - 1 - 2 91 115
AY145502.1 - 1 - 2 52 115
BC012014.1 - 1 - 2 50 115
BC037247.1 - 1 - 2 67 115
BC047565.1 - 1 - 2 119 115
GQ472228.1 - 1 - 2 119 115

View Full Product Details