Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008910.2 | NP_001008910.1 | 1 - 2 | 54 | 72 |
XM_011512067.2 | XP_011510369.1 | 1 - 2 | 55 | 72 | |
GenBank mRNA | AK292694.1 | - | 1 - 2 | 54 | 72 |
AK298471.1 | - | 1 - 2 | 42 | 72 |