Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001363.4 | NP_001354.1 | 12 - 13 | 1475 | 70 |
NR_110021.1 | 11 - 12 | 1952 | 70 | ||
NR_110022.1 | 11 - 12 | 2071 | 70 | ||
NR_110023.1 | 12 - 13 | 1845 | 70 | ||
GenBank mRNA | AF067008.1 | - | 12 - 13 | 1311 | 70 |
AK225077.1 | - | 12 - 13 | 1339 | 70 | |
BC009928.2 | - | 12 - 13 | 1453 | 70 | |
BC010015.2 | - | 12 - 13 | 1277 | 70 | |
KC954523.1 | - | 11 - 12 | 1775 | 70 | |
KC954524.1 | - | 11 - 12 | 1894 | 70 | |
KC954525.1 | - | 12 - 13 | 1668 | 70 | |
U59151.1 | - | 12 - 13 | 1331 | 70 |