Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142463.2 | NP_001135935.1 | 13 - 14 | 1551 | 68 |
NM_001288747.1 | NP_001275676.1 | 12 - 13 | 2055 | 68 | |
NM_001363.4 | NP_001354.1 | 13 - 14 | 1566 | 68 | |
NR_110021.1 | 12 - 13 | 2043 | 68 | ||
NR_110022.1 | 12 - 13 | 2162 | 68 | ||
NR_110023.1 | 13 - 14 | 1936 | 68 | ||
GenBank mRNA | AF067008.1 | - | 13 - 14 | 1402 | 68 |
AK225077.1 | - | 13 - 14 | 1430 | 68 | |
AK307672.1 | - | 13 - 14 | 1440 | 68 | |
BC009928.2 | - | 13 - 14 | 1544 | 68 | |
BC010015.2 | - | 13 - 14 | 1368 | 68 | |
JF279874.1 | - | 12 - 13 | 1878 | 68 | |
KC954523.1 | - | 12 - 13 | 1866 | 68 | |
KC954524.1 | - | 12 - 13 | 1985 | 68 | |
KC954525.1 | - | 13 - 14 | 1759 | 68 | |
U59151.1 | - | 13 - 14 | 1422 | 68 |