Assay Design



Gene Transcripts

Gene Symbol
AKR1B15
Entrez Gene ID
441282
Gene Name
aldo-keto reductase family 1 member B15
Gene Aliases
AKR1B10L, AKR1B10L, AK1R1B7, AKR1R1B7
Chromosome Location
Chr.7: 134549097 - 134579841 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080538.2 NP_001074007.2 10 - 11 1203 136
XM_011516241.2 XP_011514543.2 8 - 9 1605 136
XM_017012224.1 XP_016867713.1 8 - 9 956 136

View Full Product Details