Assay Design



Gene Transcripts

Gene Symbol
PPP6R2
Entrez Gene ID
9701
Gene Name
protein phosphatase 6 regulatory subunit 2
Gene Aliases
KIAA0685, PP6R2, SAP190, SAPS2
Chromosome Location
Chr.22: 50343317 - 50445090 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242898.1 NP_001229827.1 13 - 14 1897 231
NM_001242899.1 NP_001229828.1 13 - 14 1900 150
NM_001242900.1 NP_001229829.1 13 - 14 1897 150
NM_014678.4 NP_055493.2 13 - 14 1897 150
XM_005261960.3 XP_005262017.1 7 - 8 851 231
XM_006724431.2 XP_006724494.1 14 - 15 2008 231
XM_006724434.1 XP_006724497.1 15 - 16 2157 231
XM_006724435.2 XP_006724498.1 13 - 14 1882 150
XM_011530720.2 XP_011529022.1 12 - 13 1585 231
XM_011530721.2 XP_011529023.1 12 - 13 1582 231
XM_011530722.2 XP_011529024.1 12 - 13 1582 231
XM_011530723.2 XP_011529025.1 12 - 13 1585 231
XM_011530724.2 XP_011529026.1 12 - 13 1585 231
XM_011530726.2 XP_011529028.1 12 - 13 1585 150
XM_011530727.2 XP_011529029.1 12 - 13 1585 150
XM_011530728.2 XP_011529030.1 12 - 13 1585 150
XM_011530729.2 XP_011529031.1 12 - 13 1585 150
XM_011530730.2 XP_011529032.1 12 - 13 1582 150
XM_011530731.1 XP_011529033.1 12 - 13 1582 150
XM_011530732.2 XP_011529034.1 15 - 16 2061 231
XM_011530734.2 XP_011529036.1 14 - 15 1935 231
XM_011530735.2 XP_011529037.1 13 - 14 1885 231
XM_011530736.2 XP_011529038.1 14 - 15 2011 231
XM_011530737.2 XP_011529039.1 15 - 16 2160 231
XM_011530738.2 XP_011529040.1 14 - 15 1963 231
XM_011530739.2 XP_011529041.1 12 - 13 1625 231
XM_011530740.2 XP_011529042.1 14 - 15 2033 231
XM_017029116.1 XP_016884605.1 12 - 13 1582 150
XM_017029117.1 XP_016884606.1 16 - 17 2210 231
XM_017029118.1 XP_016884607.1 16 - 17 2162 231
XM_017029119.1 XP_016884608.1 15 - 16 2084 231
XM_017029120.1 XP_016884609.1 15 - 16 2058 231
XM_017029121.1 XP_016884610.1 13 - 14 1882 231
XM_017029122.1 XP_016884611.1 14 - 15 1960 231
XM_017029123.1 XP_016884612.1 13 - 14 1882 231
XM_017029124.1 XP_016884613.1 14 - 15 2008 231
XM_017029125.1 XP_016884614.1 13 - 14 1882 231
XM_017029126.1 XP_016884615.1 13 - 14 1885 231
XM_017029127.1 XP_016884616.1 14 - 15 1960 231
XM_017029128.1 XP_016884617.1 14 - 15 2008 231
XM_017029129.1 XP_016884618.1 13 - 14 1882 150
XM_017029130.1 XP_016884619.1 13 - 14 1882 150
XM_017029131.1 XP_016884620.1 14 - 15 2008 150
XM_017029132.1 XP_016884621.1 14 - 15 1960 150
XM_017029133.1 XP_016884622.1 13 - 14 1839 231
XM_017029134.1 XP_016884623.1 13 - 14 1836 231
XM_017029135.1 XP_016884624.1 14 - 15 1886 231
XM_017029136.1 XP_016884625.1 13 - 14 1836 231
XM_017029137.1 XP_016884626.1 13 - 14 1836 231
XM_017029138.1 XP_016884627.1 13 - 14 1836 150
GenBank mRNA AB014585.1 - 13 - 14 1869 150
AK002063.1 - 6 - 7 693 231
AK022669.1 - 3 - 4 401 150
AK302472.1 - 13 - 14 1808 231
BC000976.4 - 14 - 15 1886 231
BC006568.2 - 13 - 14 1883 150
BC032664.1 - 13 - 14 1897 150
BC041698.1 - 13 - 14 1867 150
BC052995.2 - 13 - 14 1803 150

View Full Product Details