Assay Design



Gene Transcripts

Gene Symbol
LARP4
Entrez Gene ID
113251
Gene Name
La ribonucleoprotein domain family member 4
Gene Aliases
PP13296
Chromosome Location
Chr.12: 50400809 - 50480005 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170804.1 NP_001164275.1 9 - 10 1226 129
NM_001170808.1 NP_001164279.1 9 - 10 1226 129
NM_052879.4 NP_443111.4 9 - 10 1226 129
NM_199188.2 NP_954658.2 9 - 10 1223 129
NR_033200.1 10 - 11 1275 129
XM_005268613.2 XP_005268670.1 10 - 11 1275 129
XM_011537828.2 XP_011536130.1 10 - 11 1330 129
XM_011537829.2 XP_011536131.1 10 - 11 1311 129
XM_011537830.2 XP_011536132.1 9 - 10 1158 129
XM_011537831.2 XP_011536133.1 9 - 10 1222 129
XM_011537834.2 XP_011536136.1 10 - 11 1331 129
XM_011537836.2 XP_011536138.1 10 - 11 1312 129
XM_011537838.2 XP_011536140.1 9 - 10 1070 129
XM_011537839.2 XP_011536141.1 10 - 11 1214 129
XM_011537840.2 XP_011536142.1 10 - 11 1267 129
XM_011537841.2 XP_011536143.1 9 - 10 1120 129
XM_011537844.2 XP_011536146.1 10 - 11 1331 129
XM_017018751.1 XP_016874240.1 10 - 11 1091 129
XM_017018752.1 XP_016874241.1 9 - 10 1074 129
XM_017018753.1 XP_016874242.1 9 - 10 1107 129
XM_017018754.1 XP_016874243.1 10 - 11 1257 129
XM_017018759.1 XP_016874248.1 9 - 10 1120 129
XM_017018760.1 XP_016874249.1 9 - 10 1226 129
XM_017018762.1 XP_016874251.1 9 - 10 1219 129
GenBank mRNA AF370416.1 - 9 - 10 1104 129
AK055521.1 - 7 - 8 774 129
AK125113.1 - 8 - 9 1016 129
AK291746.1 - 9 - 10 1095 129
AL832198.1 - 9 - 10 1167 129
AL833272.1 - 9 - 10 1136 129
AY004310.1 - 10 - 11 1130 129
BC063479.1 - 9 - 10 1127 129
CR936626.1 - 9 - 10 1144 129

View Full Product Details