Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004452.3 | NP_004443.3 | 8 - 9 | 1428 | 107 |
XM_011536547.2 | XP_011534849.1 | 6 - 7 | 1590 | 107 | |
XM_011536550.2 | XP_011534852.1 | 8 - 9 | 1664 | 107 | |
XM_011536553.2 | XP_011534855.1 | 6 - 7 | 1590 | 107 | |
XM_011536554.2 | XP_011534856.1 | 6 - 7 | 1590 | 107 | |
XM_017021085.1 | XP_016876574.1 | 7 - 8 | 1235 | 107 | |
XM_017021086.1 | XP_016876575.1 | 8 - 9 | 1665 | 107 | |
GenBank mRNA | AB307714.1 | - | 5 - 6 | 1057 | 107 |
AY451389.1 | - | 6 - 7 | 1132 | 107 | |
AY451390.1 | - | 6 - 7 | 1132 | 107 | |
BC131517.1 | - | 7 - 8 | 1155 | 107 | |
HQ692852.1 | - | 5 - 6 | 1057 | 107 |