Assay Design



Gene Transcripts

Gene Symbol
NEGR1
Entrez Gene ID
257194
Gene Name
neuronal growth regulator 1
Gene Aliases
DMML2433, IGLON4, KILON, Ntra
Chromosome Location
Chr.1: 71402942 - 72282850 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173808.2 NP_776169.2 1 - 2 272 114
XM_011541200.2 XP_011539502.1 1 - 2 528 114
XM_011541201.2 XP_011539503.1 1 - 2 528 114
XM_017000961.1 XP_016856450.1 1 - 2 528 114
GenBank mRNA AK092307.1 - 1 - 2 179 114
AK295481.1 - 1 - 2 170 114
BX538014.1 - 1 - 2 176 114

View Full Product Details