Assay Design
Gene Transcripts
- Gene Symbol
- AADACL2
- Entrez Gene ID
-
344752
- Gene Name
- arylacetamide deacetylase like 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.3: 151733916 - 151758016 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_207365.3 |
NP_997248.2 |
1 - 2 |
256 |
147 |
GenBank mRNA |
BC065724.1 |
- |
1 - 2 |
249 |
147 |
View Full Product Details