Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015111.1 | NP_055926.1 | 3 - 4 | 1106 | 87 |
XM_006714834.3 | XP_006714897.1 | 3 - 4 | 1136 | 87 | |
XM_011534473.1 | XP_011532775.1 | 3 - 4 | 1155 | 87 | |
XM_011534474.1 | XP_011532776.1 | 3 - 4 | 1155 | 87 | |
GenBank mRNA | AB002339.1 | - | 2 - 3 | 867 | 87 |
BC053323.1 | - | 3 - 4 | 1106 | 87 |