Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000627.3 | NP_000618.3 | 29 - 30 | 4149 | 144 |
NM_001166264.1 | NP_001159736.1 | 28 - 29 | 4023 | 144 | |
NM_001166265.1 | NP_001159737.1 | 29 - 30 | 3990 | 144 | |
NM_001166266.1 | NP_001159738.1 | 28 - 29 | 3864 | 144 | |
NM_206943.2 | NP_996826.2 | 33 - 34 | 5009 | 144 | |
XM_005264317.3 | XP_005264374.1 | 33 - 34 | 4852 | 144 | |
XM_005264318.3 | XP_005264375.1 | 32 - 33 | 4726 | 144 | |
XM_011532853.2 | XP_011531155.1 | 33 - 34 | 5185 | 144 | |
XM_011532855.2 | XP_011531157.1 | 33 - 34 | 5008 | 144 | |
XM_011532856.2 | XP_011531158.1 | 32 - 33 | 4894 | 144 | |
XM_011532857.2 | XP_011531159.1 | 32 - 33 | 4888 | 144 | |
XM_011532858.2 | XP_011531160.1 | 32 - 33 | 4888 | 144 | |
XM_011532859.2 | XP_011531161.1 | 33 - 34 | 4855 | 144 | |
XM_011532860.2 | XP_011531162.1 | 32 - 33 | 4846 | 144 | |
XM_011532861.2 | XP_011531163.1 | 31 - 32 | 4744 | 144 | |
XM_017004108.1 | XP_016859597.1 | 32 - 33 | 4891 | 144 | |
XM_017004109.1 | XP_016859598.1 | 32 - 33 | 4885 | 144 | |
XM_017004110.1 | XP_016859599.1 | 31 - 32 | 4765 | 144 | |
GenBank mRNA | AB208801.1 | - | 29 - 30 | 3948 | 144 |
AK298027.1 | - | 23 - 24 | 2718 | 144 | |
AK299237.1 | - | 11 - 12 | 1494 | 144 | |
AK299912.1 | - | 23 - 24 | 2821 | 144 | |
AK302857.1 | - | 14 - 15 | 1888 | 144 | |
AK315991.1 | - | 14 - 15 | 2052 | 144 | |
BC130289.1 | - | 29 - 30 | 4089 | 144 | |
BC144127.1 | - | 28 - 29 | 3806 | 144 | |
BC144128.1 | - | 28 - 29 | 3963 | 144 | |
BX103725.1 | - | 1 - 2 | 151 | 144 |