Assay Design



Gene Transcripts

Gene Symbol
LTBP1
Entrez Gene ID
4052
Gene Name
latent transforming growth factor beta binding protein 1
Gene Aliases
-
Chromosome Location
Chr.2: 32947129 - 33399509 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000627.3 NP_000618.3 3 - 4 861 171
NM_001166264.1 NP_001159736.1 3 - 4 861 171
NM_001166265.1 NP_001159737.1 3 - 4 861 171
NM_001166266.1 NP_001159738.1 3 - 4 861 171
NM_206943.2 NP_996826.2 7 - 8 1721 171
XM_005264317.3 XP_005264374.1 7 - 8 1723 171
XM_005264318.3 XP_005264375.1 7 - 8 1723 171
XM_011532853.2 XP_011531155.1 7 - 8 1894 171
XM_011532855.2 XP_011531157.1 7 - 8 1723 171
XM_011532856.2 XP_011531158.1 7 - 8 1723 171
XM_011532857.2 XP_011531159.1 7 - 8 1723 171
XM_011532858.2 XP_011531160.1 7 - 8 1723 171
XM_011532859.2 XP_011531161.1 7 - 8 1723 171
XM_011532860.2 XP_011531162.1 6 - 7 1555 171
XM_011532861.2 XP_011531163.1 7 - 8 1723 171
XM_011532862.2 XP_011531164.1 7 - 8 1723 171
XM_017004108.1 XP_016859597.1 7 - 8 1723 171
XM_017004109.1 XP_016859598.1 7 - 8 1723 171
XM_017004110.1 XP_016859599.1 7 - 8 1723 171
GenBank mRNA AB208801.1 - 3 - 4 819 171
AF489528.1 - 3 - 4 720 171
BC130289.1 - 3 - 4 801 171
BC144127.1 - 3 - 4 801 171
BC144128.1 - 3 - 4 801 171
M34057.1 - 3 - 4 810 171

View Full Product Details