Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270546.1 | NP_001257475.1 | 6 - 7 | 1105 | 114 |
NM_006738.5 | NP_006729.4 | 14 - 15 | 5303 | 114 | |
NM_007200.4 | NP_009131.2 | 14 - 15 | 5291 | 114 | |
GenBank mRNA | AB055890.1 | - | 15 - 16 | 5310 | 114 |
AB209414.1 | - | 3 - 4 | 590 | 114 | |
AF086293.1 | - | 3 - 4 | 336 | 114 | |
AF126008.1 | - | 6 - 7 | 945 | 114 | |
AF387101.1 | - | 13 - 14 | 5097 | 114 | |
AF406992.1 | - | 14 - 15 | 5190 | 114 | |
AK026186.1 | - | 7 - 8 | 1775 | 114 | |
AK125331.1 | - | 13 - 14 | 5267 | 114 | |
AK307618.1 | - | 8 - 9 | 1309 | 114 | |
AK309805.1 | - | 6 - 7 | 974 | 114 | |
AK315885.1 | - | 6 - 7 | 1359 | 114 | |
BC009213.2 | - | 7 - 8 | 3113 | 114 | |
BC063592.1 | - | 1 - 2 | 78 | 114 | |
BC089394.1 | - | 2 - 3 | 232 | 114 | |
M90360.1 | - | 8 - 9 | 2838 | 114 |