Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270546.1 | NP_001257475.1 | 14 - 15 | 2002 | 135 |
NM_006738.5 | NP_006729.4 | 22 - 23 | 6203 | 135 | |
NM_007200.4 | NP_009131.2 | 22 - 23 | 6191 | 135 | |
GenBank mRNA | AB055890.1 | - | 23 - 24 | 6210 | 135 |
AB209414.1 | - | 11 - 12 | 1484 | 135 | |
AF126008.1 | - | 14 - 15 | 1845 | 135 | |
AF387101.1 | - | 21 - 22 | 5997 | 135 | |
AF406992.1 | - | 22 - 23 | 6090 | 135 | |
AK309805.1 | - | 14 - 15 | 1871 | 135 | |
BC063592.1 | - | 9 - 10 | 978 | 135 | |
BC140862.1 | - | 7 - 8 | 814 | 135 | |
BC171798.1 | - | 7 - 8 | 814 | 135 |