Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167580.1 | NP_001161052.1 | 12 - 12 | 9951 | 124 |
NM_001323031.1 | NP_001309960.1 | 13 - 13 | 10484 | 124 | |
NM_001323032.1 | NP_001309961.1 | 13 - 13 | 10793 | 124 | |
NM_001323033.1 | NP_001309962.1 | 11 - 11 | 10460 | 124 | |
NM_001323034.1 | NP_001309963.1 | 12 - 12 | 10592 | 124 | |
NM_001323036.1 | NP_001309965.1 | 12 - 12 | 9770 | 124 | |
NM_001323037.1 | NP_001309966.1 | 14 - 14 | 10700 | 124 | |
NM_001323038.1 | NP_001309967.1 | 13 - 13 | 10612 | 124 | |
NM_001323039.1 | NP_001309968.1 | 13 - 13 | 10885 | 124 | |
NM_001323040.1 | NP_001309969.1 | 13 - 13 | 10131 | 124 | |
NM_014848.5 | NP_055663.1 | 14 - 14 | 10881 | 124 | |
XM_005254998.3 | XP_005255055.1 | 13 - 13 | 10546 | 124 | |
XM_017022761.1 | XP_016878250.1 | 13 - 13 | 10497 | 124 | |
XM_017022762.1 | XP_016878251.1 | 13 - 13 | 10632 | 124 | |
GenBank mRNA | AL109781.1 | - | 1 - 1 | 558 | 124 |