Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178439.4 | NP_848526.1 | 4 - 5 | 866 | 116 |
XM_011533033.1 | XP_011531335.1 | 4 - 5 | 830 | 116 | |
GenBank mRNA | AK023119.1 | - | 4 - 5 | 772 | 116 |
AK023261.1 | - | 3 - 4 | 208 | 116 | |
AK024042.1 | - | 4 - 5 | 830 | 116 | |
BC007420.2 | - | 4 - 5 | 786 | 116 |