Assay Design



Gene Transcripts

Gene Symbol
ARNTL
Entrez Gene ID
406
Gene Name
aryl hydrocarbon receptor nuclear translocator like
Gene Aliases
BMAL1, BMAL1c, JAP3, MOP3, PASD3, TIC, bHLHe5
Chromosome Location
Chr.11: 13276613 - 13387268 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030272.2 NP_001025443.1 14 - 15 1732 108
NM_001030273.2 NP_001025444.1 15 - 16 1803 108
NM_001178.5 NP_001169.3 15 - 16 1787 108
NM_001297719.1 NP_001284648.1 15 - 16 1790 108
NM_001297722.1 NP_001284651.1 14 - 15 1735 108
NM_001297724.1 NP_001284653.1 15 - 16 1799 108
XM_006718233.3 XP_006718296.1 14 - 15 1677 108
XM_006718234.1 XP_006718297.1 15 - 16 1743 108
XM_011520105.2 XP_011518407.2 13 - 14 1838 108
XM_011520107.2 XP_011518409.2 13 - 14 1833 108
XM_011520108.1 XP_011518410.1 15 - 16 1783 108
XM_011520109.1 XP_011518411.1 13 - 14 1538 108
XM_011520110.1 XP_011518412.1 16 - 17 1798 108
XM_011520111.1 XP_011518413.1 14 - 15 1613 108
XM_011520112.2 XP_011518414.1 16 - 17 1798 108
XM_011520113.1 XP_011518415.1 16 - 17 1795 108
XM_017017738.1 XP_016873227.1 13 - 14 1839 108
XM_017017739.1 XP_016873228.1 13 - 14 1835 108
XM_017017740.1 XP_016873229.1 14 - 15 1674 108
XM_017017741.1 XP_016873230.1 13 - 14 1535 108
XM_017017742.1 XP_016873231.1 13 - 14 1538 108
XM_017017743.1 XP_016873232.1 13 - 14 1535 108
XM_017017744.1 XP_016873233.1 16 - 17 1795 108
XM_017017745.1 XP_016873234.1 15 - 16 1740 108
XM_017017746.1 XP_016873235.1 15 - 16 1753 108
XM_017017747.1 XP_016873236.1 14 - 15 1613 108
XM_017017748.1 XP_016873237.1 14 - 15 1667 108
GenBank mRNA AB000812.1 - 12 - 13 1371 108
AF044288.1 - 15 - 16 1700 108
AK095749.1 - 15 - 16 1688 108
AK128317.1 - 1 - 2 90 108
AK291510.1 - 14 - 15 1648 108
AK315174.1 - 15 - 16 1728 108
BC016674.1 - 12 - 13 1334 108
BC031214.1 - 15 - 16 1631 108
BC041129.1 - 15 - 16 1711 108
D89722.1 - 12 - 13 1347 108
U51627.1 - 9 - 10 1329 108
U60415.1 - 14 - 15 1634 108

View Full Product Details