Assay Design



Gene Transcripts

Gene Symbol
ARNTL
Entrez Gene ID
406
Gene Name
aryl hydrocarbon receptor nuclear translocator like
Gene Aliases
BMAL1, BMAL1c, JAP3, MOP3, PASD3, TIC, bHLHe5
Chromosome Location
Chr.11: 13276613 - 13387268 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030272.2 NP_001025443.1 10 - 11 1078 150
NM_001030273.2 NP_001025444.1 11 - 12 1149 150
NM_001178.5 NP_001169.3 11 - 12 1133 150
NM_001297719.1 NP_001284648.1 11 - 12 1133 150
NM_001297722.1 NP_001284651.1 10 - 11 1078 150
NM_001297724.1 NP_001284653.1 11 - 12 1142 150
XM_006718233.3 XP_006718296.1 10 - 11 1020 150
XM_006718234.1 XP_006718297.1 11 - 12 1086 150
XM_011520105.2 XP_011518407.2 9 - 10 1184 150
XM_011520107.2 XP_011518409.2 9 - 10 1179 150
XM_011520108.1 XP_011518410.1 11 - 12 1126 150
XM_011520109.1 XP_011518411.1 9 - 10 881 150
XM_011520110.1 XP_011518412.1 12 - 13 1141 150
XM_011520111.1 XP_011518413.1 10 - 11 956 150
XM_011520112.2 XP_011518414.1 12 - 13 1141 150
XM_011520113.1 XP_011518415.1 12 - 13 1141 150
XM_017017738.1 XP_016873227.1 9 - 10 1182 150
XM_017017739.1 XP_016873228.1 9 - 10 1178 150
XM_017017740.1 XP_016873229.1 10 - 11 1020 150
XM_017017741.1 XP_016873230.1 9 - 10 881 150
XM_017017742.1 XP_016873231.1 9 - 10 881 150
XM_017017743.1 XP_016873232.1 9 - 10 881 150
XM_017017744.1 XP_016873233.1 12 - 13 1141 150
XM_017017745.1 XP_016873234.1 11 - 12 1086 150
XM_017017746.1 XP_016873235.1 11 - 12 1096 150
XM_017017747.1 XP_016873236.1 10 - 11 956 150
XM_017017748.1 XP_016873237.1 10 - 11 1013 150
GenBank mRNA AB000812.1 - 8 - 9 714 150
AB000815.1 - 9 - 10 785 150
AB000816.1 - 10 - 11 1125 150
AF044288.1 - 11 - 12 1043 150
AK095749.1 - 11 - 12 1034 150
AK291510.1 - 10 - 11 991 150
AK315174.1 - 11 - 12 1071 150
BC016674.1 - 8 - 9 677 150
BC031214.1 - 11 - 12 974 150
BC041129.1 - 11 - 12 1057 150
D89722.1 - 8 - 9 690 150
U51627.1 - 5 - 6 672 150
U60415.1 - 10 - 11 977 150

View Full Product Details