Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002344.5 | NP_002335.2 | 6 - 7 | 991 | 129 |
XM_011521556.2 | XP_011519858.1 | 6 - 7 | 991 | 129 | |
XM_017022181.1 | XP_016877670.1 | 6 - 7 | 991 | 129 | |
XM_017022182.1 | XP_016877671.1 | 6 - 7 | 991 | 129 | |
XM_017022183.1 | XP_016877672.1 | 6 - 7 | 991 | 129 | |
GenBank mRNA | AK314006.1 | - | 6 - 7 | 958 | 129 |
BC045607.1 | - | 6 - 7 | 964 | 129 | |
D16105.1 | - | 6 - 7 | 910 | 129 |