Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170803.1 | NP_001164274.1 | 1 - 2 | 219 | 148 |
NM_001170804.1 | NP_001164275.1 | 1 - 2 | 219 | 148 | |
NM_001170808.1 | NP_001164279.1 | 1 - 2 | 219 | 148 | |
NM_052879.4 | NP_443111.4 | 1 - 2 | 219 | 148 | |
NM_199190.2 | NP_954660.1 | 1 - 2 | 219 | 148 | |
XM_011537830.2 | XP_011536132.1 | 1 - 2 | 133 | 148 | |
XM_017018750.1 | XP_016874239.1 | 1 - 2 | 127 | 148 | |
XM_017018760.1 | XP_016874249.1 | 1 - 2 | 219 | 148 | |
XM_017018761.1 | XP_016874250.1 | 1 - 2 | 212 | 148 | |
XM_017018762.1 | XP_016874251.1 | 1 - 2 | 212 | 148 | |
GenBank mRNA | AL832198.1 | - | 1 - 2 | 162 | 148 |
AL833272.1 | - | 1 - 2 | 129 | 148 | |
BC022377.1 | - | 1 - 2 | 121 | 148 | |
BC063479.1 | - | 1 - 2 | 120 | 148 | |
CR936626.1 | - | 1 - 2 | 137 | 148 |