Assay Design



Gene Transcripts

Gene Symbol
LARP4
Entrez Gene ID
113251
Gene Name
La ribonucleoprotein domain family member 4
Gene Aliases
PP13296
Chromosome Location
Chr.12: 50400809 - 50480005 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_052879.4 NP_443111.4 11 - 12 1532 133
NM_199188.2 NP_954658.2 11 - 12 1529 133
NM_199190.2 NP_954660.1 10 - 11 1319 133
XM_005268613.2 XP_005268670.1 12 - 13 1581 133
XM_011537828.2 XP_011536130.1 12 - 13 1636 133
XM_011537829.2 XP_011536131.1 12 - 13 1617 133
XM_011537830.2 XP_011536132.1 11 - 12 1464 133
XM_011537831.2 XP_011536133.1 11 - 12 1528 133
XM_011537833.2 XP_011536135.1 11 - 12 1423 133
XM_011537835.2 XP_011536137.1 11 - 12 1404 133
XM_011537838.2 XP_011536140.1 11 - 12 1376 133
XM_011537839.2 XP_011536141.1 12 - 13 1520 133
XM_011537840.2 XP_011536142.1 12 - 13 1573 133
XM_011537841.2 XP_011536143.1 11 - 12 1426 133
XM_011537844.2 XP_011536146.1 12 - 13 1637 133
XM_017018750.1 XP_016874239.1 10 - 11 1245 133
XM_017018751.1 XP_016874240.1 12 - 13 1397 133
XM_017018752.1 XP_016874241.1 11 - 12 1380 133
XM_017018753.1 XP_016874242.1 11 - 12 1413 133
XM_017018754.1 XP_016874243.1 12 - 13 1563 133
XM_017018755.1 XP_016874244.1 10 - 11 1230 133
XM_017018756.1 XP_016874245.1 11 - 12 1282 133
XM_017018757.1 XP_016874246.1 12 - 13 1452 133
XM_017018758.1 XP_016874247.1 11 - 12 1388 133
XM_017018760.1 XP_016874249.1 11 - 12 1532 133
XM_017018761.1 XP_016874250.1 10 - 11 1312 133
GenBank mRNA AF370416.1 - 11 - 12 1410 133
AK055521.1 - 9 - 10 1080 133
AK125113.1 - 10 - 11 1322 133
AK291746.1 - 11 - 12 1401 133
AL833272.1 - 11 - 12 1442 133
CR936626.1 - 11 - 12 1450 133

View Full Product Details