Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199155.1 | NP_001186084.1 | 13 - 14 | 1862 | 100 |
NM_001199156.1 | NP_001186085.1 | 12 - 13 | 1535 | 100 | |
NM_001199157.1 | NP_001186086.1 | 12 - 13 | 1622 | 100 | |
NM_001199158.1 | NP_001186087.1 | 11 - 12 | 1445 | 100 | |
NM_007067.4 | NP_008998.1 | 14 - 15 | 1952 | 100 | |
XM_011524235.2 | XP_011522537.1 | 8 - 9 | 1002 | 100 | |
GenBank mRNA | AF074606.1 | - | 14 - 15 | 1778 | 100 |
AF140360.1 | - | 14 - 15 | 1875 | 100 | |
AF217502.1 | - | 14 - 15 | 1860 | 100 | |
AK023890.1 | - | 14 - 15 | 1883 | 100 | |
AK122932.1 | - | 14 - 15 | 2318 | 100 | |
AK293976.1 | - | 12 - 13 | 1473 | 100 | |
AK294014.1 | - | 12 - 13 | 1559 | 100 | |
AK294052.1 | - | 11 - 12 | 1371 | 100 | |
AK294367.1 | - | 9 - 10 | 1061 | 100 | |
AK294602.1 | - | 7 - 8 | 974 | 100 | |
AK294836.1 | - | 14 - 15 | 1767 | 100 | |
AK295914.1 | - | 8 - 9 | 1107 | 100 | |
AK297212.1 | - | 13 - 14 | 1944 | 100 | |
BC032640.2 | - | 14 - 15 | 1862 | 100 |