Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018008.1 | NP_001018008.1 | 7 - 8 | 806 | 150 |
XM_005254645.2 | XP_005254702.2 | 8 - 9 | 1409 | 150 | |
XM_005254653.2 | XP_005254710.1 | 7 - 8 | 792 | 150 | |
XM_017022539.1 | XP_016878028.1 | 8 - 9 | 1409 | 150 | |
GenBank mRNA | AB209041.1 | - | 7 - 8 | 843 | 150 |
BC050473.1 | - | 7 - 8 | 793 | 150 | |
BC053545.1 | - | 7 - 8 | 804 | 150 |