Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181787.2 | NP_861452.2 | 10 - 11 | 1230 | 149 |
XM_005250894.4 | XP_005250951.2 | 10 - 11 | 1443 | 149 | |
XM_005250895.3 | XP_005250952.1 | 9 - 10 | 1111 | 149 | |
XM_011516984.2 | XP_011515286.1 | 9 - 10 | 1110 | 149 | |
XM_017013351.1 | XP_016868840.1 | 9 - 10 | 1104 | 149 | |
XM_017013352.1 | XP_016868841.1 | 8 - 9 | 1181 | 149 | |
XM_017013354.1 | XP_016868843.1 | 9 - 10 | 1136 | 149 | |
XM_017013355.1 | XP_016868844.1 | 9 - 10 | 1129 | 149 | |
XM_017013356.1 | XP_016868845.1 | 8 - 9 | 982 | 149 | |
XM_017013357.1 | XP_016868846.1 | 8 - 9 | 970 | 149 | |
GenBank mRNA | AK123618.1 | - | 2 - 3 | 225 | 149 |
AK123682.1 | - | 10 - 11 | 1184 | 149 | |
AL832859.1 | - | 5 - 6 | 545 | 149 | |
BC110870.1 | - | 10 - 11 | 1158 | 149 | |
BC126193.1 | - | 10 - 11 | 1126 | 149 | |
BC130576.1 | - | 10 - 11 | 1126 | 149 | |
BX538048.1 | - | 10 - 11 | 1178 | 149 | |
BX538174.1 | - | 10 - 11 | 1182 | 149 |