Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181787.2 | NP_861452.2 | 11 - 12 | 1291 | 83 |
XM_005250894.4 | XP_005250951.2 | 11 - 12 | 1504 | 83 | |
XM_005250895.3 | XP_005250952.1 | 10 - 11 | 1172 | 83 | |
XM_011516983.2 | XP_011515285.1 | 10 - 11 | 1198 | 83 | |
XM_011516984.2 | XP_011515286.1 | 10 - 11 | 1171 | 83 | |
XM_017013351.1 | XP_016868840.1 | 10 - 11 | 1165 | 83 | |
XM_017013352.1 | XP_016868841.1 | 9 - 10 | 1242 | 83 | |
XM_017013353.1 | XP_016868842.1 | 9 - 10 | 1066 | 83 | |
XM_017013354.1 | XP_016868843.1 | 10 - 11 | 1197 | 83 | |
XM_017013355.1 | XP_016868844.1 | 10 - 11 | 1190 | 83 | |
XM_017013356.1 | XP_016868845.1 | 9 - 10 | 1043 | 83 | |
XM_017013357.1 | XP_016868846.1 | 9 - 10 | 1031 | 83 | |
GenBank mRNA | AK123618.1 | - | 3 - 4 | 286 | 83 |
AK123682.1 | - | 11 - 12 | 1245 | 83 | |
AL832859.1 | - | 6 - 7 | 606 | 83 | |
BC110870.1 | - | 11 - 12 | 1219 | 83 | |
BC126193.1 | - | 11 - 12 | 1187 | 83 | |
BC130576.1 | - | 11 - 12 | 1187 | 83 | |
BX538048.1 | - | 11 - 12 | 1239 | 83 | |
BX538174.1 | - | 11 - 12 | 1243 | 83 |