Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181787.2 | NP_861452.2 | 16 - 17 | 1840 | 131 |
XM_005250894.4 | XP_005250951.2 | 16 - 17 | 2053 | 131 | |
XM_005250895.3 | XP_005250952.1 | 15 - 16 | 1721 | 131 | |
XM_011516983.2 | XP_011515285.1 | 15 - 16 | 1747 | 131 | |
XM_011516984.2 | XP_011515286.1 | 15 - 16 | 1720 | 131 | |
XM_017013351.1 | XP_016868840.1 | 15 - 16 | 1714 | 131 | |
XM_017013352.1 | XP_016868841.1 | 14 - 15 | 1791 | 131 | |
XM_017013353.1 | XP_016868842.1 | 14 - 15 | 1615 | 131 | |
XM_017013354.1 | XP_016868843.1 | 15 - 16 | 1746 | 131 | |
XM_017013355.1 | XP_016868844.1 | 15 - 16 | 1739 | 131 | |
XM_017013356.1 | XP_016868845.1 | 14 - 15 | 1592 | 131 | |
XM_017013357.1 | XP_016868846.1 | 14 - 15 | 1580 | 131 | |
GenBank mRNA | AK123618.1 | - | 8 - 9 | 835 | 131 |
AK123682.1 | - | 16 - 17 | 1794 | 131 | |
AL832859.1 | - | 10 - 11 | 1041 | 131 | |
BC110870.1 | - | 16 - 17 | 1768 | 131 | |
BC126193.1 | - | 16 - 17 | 1736 | 131 | |
BC130576.1 | - | 16 - 17 | 1736 | 131 | |
BX538048.1 | - | 16 - 17 | 1788 | 131 | |
BX538174.1 | - | 16 - 17 | 1792 | 131 |