Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181787.2 | NP_861452.2 | 17 - 18 | 1975 | 140 |
XM_005250894.4 | XP_005250951.2 | 17 - 18 | 2188 | 140 | |
XM_005250895.3 | XP_005250952.1 | 16 - 17 | 1856 | 140 | |
XM_011516983.2 | XP_011515285.1 | 16 - 17 | 1882 | 140 | |
XM_011516984.2 | XP_011515286.1 | 16 - 17 | 1855 | 140 | |
XM_017013351.1 | XP_016868840.1 | 16 - 17 | 1849 | 140 | |
XM_017013352.1 | XP_016868841.1 | 15 - 16 | 1926 | 140 | |
XM_017013353.1 | XP_016868842.1 | 15 - 16 | 1750 | 140 | |
XM_017013354.1 | XP_016868843.1 | 16 - 17 | 1881 | 140 | |
XM_017013355.1 | XP_016868844.1 | 16 - 17 | 1874 | 140 | |
XM_017013356.1 | XP_016868845.1 | 15 - 16 | 1727 | 140 | |
XM_017013357.1 | XP_016868846.1 | 15 - 16 | 1715 | 140 | |
GenBank mRNA | AK123618.1 | - | 9 - 10 | 970 | 140 |
AK123682.1 | - | 17 - 18 | 1929 | 140 | |
AL832859.1 | - | 11 - 12 | 1176 | 140 | |
BC110870.1 | - | 17 - 18 | 1903 | 140 | |
BC126193.1 | - | 17 - 18 | 1871 | 140 | |
BC130576.1 | - | 17 - 18 | 1871 | 140 | |
BX538048.1 | - | 17 - 18 | 1923 | 140 | |
BX538174.1 | - | 17 - 18 | 1927 | 140 |