Assay Design
Gene Transcripts
- Gene Symbol
- GRIK1-AS1
- Entrez Gene ID
-
642976
- Gene Name
- GRIK1 antisense RNA 1
- Gene Aliases
-
C21orf9, GRIK1-AS, GRIK1AS, NCRNA00110
- Chromosome Location
-
Chr.21: 29748175 - 29764006 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_027021.1 |
|
2 - 3 |
442 |
143 |
GenBank mRNA |
AY077697.1 |
- |
1 - 2 |
212 |
143 |
BG187681.1 |
- |
2 - 3 |
447 |
143 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
GRIK1 antisense RNA 2 |
GRIK1-AS2 |
C21orf41, NCRNA00258 |
-118.1kb
|
Sense |
Chr. - 29596040 - 29630747 on Build GRCh38 |
Downstream |
claudin 17 |
CLDN17 |
|
406.9kb
|
Anti-sense |
Chr. - 30165923 - 30166653 on Build GRCh38 |
View Full Product Details