Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002834.3 | NP_002825.3 | 10 - 11 | 1598 | 78 |
NM_080601.1 | NP_542168.1 | 10 - 11 | 1598 | 78 | |
XM_017019722.1 | XP_016875211.1 | 10 - 11 | 1415 | 78 | |
GenBank mRNA | AK289854.1 | - | 10 - 11 | 1376 | 78 |
BC008692.1 | - | 10 - 11 | 1421 | 78 | |
BT007106.1 | - | 9 - 10 | 1218 | 78 | |
D13540.1 | - | 10 - 11 | 1374 | 78 | |
L03535.1 | - | 10 - 11 | 1331 | 78 | |
L07527.1 | - | 10 - 11 | 1371 | 78 | |
L08807.1 | - | 10 - 11 | 1416 | 78 | |
S78088.1 | - | 1 - 2 | 114 | 78 | |
X70766.1 | - | 10 - 11 | 1347 | 78 |