Assay Design



Gene Transcripts

Gene Symbol
DNAJC4
Entrez Gene ID
3338
Gene Name
DnaJ heat shock protein family (Hsp40) member C4
Gene Aliases
DANJC4, HSPF2, MCG18
Chromosome Location
Chr.11: 64230281 - 64234281 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307980.1 NP_001294909.1 3 - 4 790 149
GenBank mRNA BC032137.2 - 3 - 4 464 149

View Full Product Details