Assay Design
Gene Transcripts
- Gene Symbol
- CDC25B
- Entrez Gene ID
-
994
- Gene Name
- cell division cycle 25B
- Gene Aliases
-
-
- Chromosome Location
-
Chr.20: 3786772 - 3806121 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_136335.1 |
|
11 - 12 |
1138 |
150 |
NR_136336.1 |
|
11 - 12 |
1365 |
150 |
View Full Product Details