Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001173988.1 | NP_001167459.1 | 8 - 9 | 1083 | 103 |
NM_024718.4 | NP_078994.3 | 8 - 9 | 1080 | 103 | |
GenBank mRNA | AK000963.1 | - | 5 - 6 | 486 | 103 |
AK001184.1 | - | 7 - 8 | 667 | 103 | |
AK027586.1 | - | 5 - 6 | 501 | 103 | |
AK094382.1 | - | 8 - 9 | 1403 | 103 | |
AK128154.2 | - | 8 - 9 | 1344 | 103 | |
AK307431.1 | - | 9 - 10 | 934 | 103 | |
BC002945.2 | - | 8 - 9 | 843 | 103 | |
BC018960.2 | - | 8 - 9 | 1026 | 103 | |
BC021095.1 | - | 8 - 9 | 784 | 103 | |
BC035786.1 | - | 8 - 9 | 1038 | 103 | |
DQ141240.1 | - | 8 - 9 | 805 | 103 | |
EF156752.1 | - | 8 - 9 | 807 | 103 | |
EF156753.1 | - | 8 - 9 | 806 | 103 |